accessibility menu, dialog, popup

UserName

H2NSOONO

CAS RN 68291-97-4

IUPAC Name: (1,2-benzoxazol-3-yl)methanesulfonamide
Synonyms: zonisamide zonisamide (product) elan brand of zonisamide zonisamidum exceglan zonisamida zonisamide preparation excegram excegran zonisamide monosodium
Molecular Weight (g/mol): 212.22
Molecular Formula: C8H8N2O3S
InChi Key: UBQNRHZMVUUOMG-UHFFFAOYSA-N
SMILES: NS(=O)(=O)CC1=NOC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (8)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results