accessibility menu, dialog, popup

UserName

CAS RN 74103-07-4

IUPAC Name: 2-amino-2-(hydroxymethyl)propane-1,3-diol; 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid
Synonyms: ketorolac; tris buffer
Molecular Weight (g/mol): 376.41
Molecular Formula: C19H24N2O6
InChi Key: BWHLPLXXIDYSNW-UHFFFAOYNA-N
SMILES: NC(CO)(CO)CO.OC(=O)C1CCN2C1=CC=C2C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (8)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results