accessibility menu, dialog, popup

UserName

CAS RN 850-52-2

H3COHOH2C

CAS RN 850-52-2

IUPAC Name: 1-hydroxy-11a-methyl-1-(prop-2-en-1-yl)-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: altrenogest
Molecular Weight (g/mol): 310.44
Molecular Formula: C21H26O2
InChi Key: VWAUPFMBXBWEQY-UHFFFAOYNA-N
SMILES: CC12C=CC3=C4CCC(=O)C=C4CCC3C1CCC2(O)CC=C
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results