CAS RN 867017-68-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tocris Bioscience™ TC-S 7001
Potent and highly selective ROCK inhibitor; orally active
2
ROCK-IN-2, MedChemExpress
MedChemExpress ROCK-IN-2 (Azaindole 1; TC-S 7001) is an orally active and ATP-competitive ROCK inhibitor with IC50s of 0.6 and 1.1 nM for human ROCK-1 and ROCK-2, respectively.
3