nginx
Dry Block Incubators
Dry Block Incubators
Dry block incubators (also known as dry baths or heating blocks) deliver consistent, contamination-free heating for tubes, vials, and plates—without water or messy maintenance. Choose from interchangeable block formats and digital controls to support workflows like sample prep, enzyme reactions, incubation steps, and temperature holding. Explore trusted options from Eppendorf, Thermo Scientific, Fisherbrand, Grant Instruments, Ohaus, and IKA for reliable performance and repeatable results.
Eppendorf™ ThermoMixer™ C Accessory: Smartblock™ Thermoblocks
Maintain accurate uniform temperatures with these thermoblocks.
Dry Block Incubators
Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters
Interchangeable modular blocks, configurable for a range of applications, increase lab versatility.
Dry Block Incubators
Eppendorf™ ThermoMixer™ C Accessory, Smartblock™ Thermoblock
Maintain accurate uniform temperatures with these thermoblocks. Eppendorf™ Smartblock™ Thermoblocks are compatible with Eppendorf ThermoMixer™ C, and support a broad range of applications.
Dry Block Incubators
FAQ
A dry block incubator (dry bath) heats samples in metal blocks for consistent, water-free temperature control. It’s used for tube incubation, enzyme reactions, sample prep, and temperature holding. Brands like Eppendorf, Thermo Scientific, Fisherbrand, Grant Instruments, Ohaus, and IKA offer blocks for common tube and plate formats.
Choose a block that matches your vessel type and throughput: 0.2mL PCR tubes/strips, 0.5–2.0mL microtubes, 5–50mL conicals, or microplates/deepwell plates. Interchangeable block systems (for example, Eppendorf SmartBlocks) help you switch formats without changing the base unit.
Look for stable temperature control, uniform heat transfer, and clear digital settings with timers. Digital dry baths from Thermo Scientific, Fisherbrand, and Grant Instruments commonly support precise setpoints and easy monitoring. Pairing the correct block and using lids or covers can reduce evaporation and improve consistency across samples.
Dry baths avoid water handling, reduce spill and contamination risk, and typically require less cleaning. They’re also compact and fast to set up, making them ideal for routine incubation steps. Many labs prefer dry block incubators for tube-based workflows where repeatable heating and a clean bench footprint matter most.
Use a heated lid or block cover when available, and select blocks designed for tight contact with your tube type. Shorter incubation times and correct fill volumes also help. Many accessories across Eppendorf, Thermo Scientific, and Grant Instruments setups are designed to improve sealing and reduce condensation or sample loss.
Dry block incubators are widely used in molecular biology, clinical and QC labs, and general sample prep areas. They’re common for PCR prep steps, enzyme incubations, resuspension, temperature holding, and gentle heat-assisted workflows. Compact options from Ohaus and IKA are popular where bench space is limited.
Top Brands