missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lipoyl synthase, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Lipoyl synthase, mitochondrial, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001265519.1 |
---|---|
Gene ID (Entrez) | 11019 |
Name | Lipoyl synthase, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16012360
|
enQuireBio™
QP6307-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16032360
|
enQuireBio™
QP6307-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16022360
|
enQuireBio™
QP6307-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16042360
|
enQuireBio™
QP6307-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052360
|
enQuireBio™
QP6307-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16062360
|
enQuireBio™
QP6307-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001265519.1 | |
Lipoyl synthase, mitochondrial Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
11019 | |
Research Use Only | |
LIAS | |
Recombinant Protein | |
E. coli | |
LSSLPDKKKE LLQNGPDLQD FVSGDLADRS TWDEYKGNLK RQKGERLRLP PWLKTEIPMG KNYNKLKNTL RNLNLHTVCE EARCPNIGEC WGGGEYATAT ATIMLMGDTC TRGCRFCSVK TARNPPPLDA SEPYNTAKAI AEWGLDYVVL TSVDRDDMPD GGAEHIAKTV SYLKERNPKI LVECLTPDFR GDLKAIEKVA LSGLDVYAHN VETVPELQSK VRDPRANFDQ SLRVLKHAKK VQPDVISKTS IMLGLGENDE QVYATMKALR EADVDCLTLG QYMQPTRRHL KVEEYITPEK FKYWEKVGNE LGFHYTASGP LVRSSYKAGE FFLKNLVAKR KTKDL | |
Greater than 90% as determined by SDS-PAGE. |