missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque IL-15-S48AA Protein
A DNA sequence encoding the Macaca mulatta (Rhesus macaque) IL-15-S48AA, was expressed in the hosts and tags indicated.
352.00 € - 3175.00 €
Specifications
Accession Number | NP_001038196.1 |
---|---|
Gene ID (Entrez) | 699616 |
Name | IL-15-S48AA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16037851
|
enQuireBio™
QP9208-YE-100UG |
100 μg |
1050.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077851
|
enQuireBio™
QP9208-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047851
|
enQuireBio™
QP9208-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067851
|
enQuireBio™
QP9208-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087851
|
enQuireBio™
QP9208-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057851
|
enQuireBio™
QP9208-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001038196.1 | |
IL-15-S48AA Protein | |
Not determined. | |
Full Length | |
Macaque | |
His | |
Tris-based buffer, 50% glycerol |
699616 | |
Research Use Only | |
IL15 | |
Recombinant Protein | |
Yeast | |
NWVNVISDLK KIEDLIQSMH IDATLYTESD VHPSCKVTAM KCFLLELQVI SHESGDTDIH DTVENLIILA NNILSSNGNI TESGCKECEE LEEKNIKEFL QSFVHIVQMF INTS | |
Greater than 90% as determined by SDS-PAGE. |