missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Acetylcholine receptor subunit alpha Protein
A DNA sequence encoding the Mus musculus (Mouse) Acetylcholine receptor subunit alpha, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_031415.2 |
---|---|
Gene ID (Entrez) | 11435 |
Name | Acetylcholine receptor subunit alpha Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15958099
|
enQuireBio™
QP5833-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978099
|
enQuireBio™
QP5833-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968099
|
enQuireBio™
QP5833-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15988099
|
enQuireBio™
QP5833-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15998099
|
enQuireBio™
QP5833-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15908109
|
enQuireBio™
QP5833-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_031415.2 | |
Acetylcholine receptor subunit alpha Protein | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
11435 | |
Research Use Only | |
Chrna1 | |
Recombinant Protein | |
E. coli | |
SEHETRLVAK LFEDYSSVVR PVEDHREIVQ VTVGLQLIQL INVDEVNQIV TTNVRLKQQW VDYNLKWNPD DYGGVKKIHI PSEKIWRPDV VLYNNADGDF AIVKFTKVLL DYTGHITWTP PAIFKSYCEI IVTHFPFDEQ NCSMKLGTWT YDGSVVAINP ESDQPDLSNF MESGEWVIKE ARGWKHWVFY SCCPTTPYLD ITYHFVMQRL | |
Greater than 80% as determined by SDS-PAGE. |