missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Mucosal addressin cell adhesion molecule 1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Mucosal addressin cell adhesion molecule 1, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_062190.1 |
---|---|
Gene ID (Entrez) | 54266 |
Name | Mucosal addressin cell adhesion molecule 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16032520
|
enQuireBio™
QP6331-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16052520
|
enQuireBio™
QP6331-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16042520
|
enQuireBio™
QP6331-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16062520
|
enQuireBio™
QP6331-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072520
|
enQuireBio™
QP6331-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16082520
|
enQuireBio™
QP6331-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_062190.1 | |
Mucosal addressin cell adhesion molecule 1 Protein | |
Not determined. | |
Extracellular Domain | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
54266 | |
Research Use Only | |
Madcam1 | |
Recombinant Protein | |
E. coli | |
QSFQVNPPEP EVAVAMGTSL QINCSMSCDK DIARVHWHGL DTNLGNVQTL PGSRVLSVRG MLSDTGTRVC VGSCGSRSFQ HSVKILVYAF PDQLEVTPEF LVPGRDQVVS CTAHNIWPAG PDSLSFALLR GEQSLEGAQA LETEQEEEMQ ETEGTPLFQV TQRWLLPSLG TPALPALYCQ VTMQLPKLVL THRRKIPVLQ SQTSPEPPST TSAKPYILTS SHTTKAVSTG LSSVALPSTP LSSEGPCYPE IHQNPEADWE LLCEASCGSG VTVHWTLAPG DLAAYHKREA GAQAWLSVLP LGPIPEGWFQ CRMDPGGQVT SLYVTGQVIP NPSS | |
Greater than 90% as determined by SDS-PAGE. |