CAS RN 1414963-82-8
CAS RN 1414963-82-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Tocris Bioscience™ NK 252
Nrf2 activator
2
NK-252, MedChemExpress
MedChemExpress NK-252 is a potential Nrf2 activator, which exhibits a great Nrf2-activating potential.
3
4