accessibility menu, dialog, popup

UserName

CAS RN 360-97-4

H2NONHNH2N

CAS RN 360-97-4

IUPAC Name: 4-amino-1H-imidazole-5-carboxamide
Synonyms: 4-aminoimidazole-5-carboxamide
Molecular Weight (g/mol): 126.12
Molecular Formula: C4H6N4O
InChi Key: DVNYTAVYBRSTGK-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(N)N=CN1
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results