accessibility menu, dialog, popup

UserName

CAS RN 494-52-0

(S)NHN

CAS RN 494-52-0

IUPAC Name: 3-[(2S)-piperidin-2-yl]pyridine
Synonyms: (-)-anabasine caswell no. 051 anabasin anabazin anabasine (substance) neonicotine epa pesticide chemical code 006001 (S)-anabasine (-)-2-(3'-pyridyl)piperidine 2-(3'-pyridyl) piperidine
Molecular Weight (g/mol): 162.24
Molecular Formula: C10H14N2
InChi Key: MTXSIJUGVMTTMU-JTQLQIEISA-N
SMILES: C1CC[C@H](NC1)C1=CC=CN=C1
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Color 
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results