accessibility menu, dialog, popup

UserName

CAS RN 536-43-6

HClH3COON

CAS RN 536-43-6

IUPAC Name: hydrogen 1-(4-butoxyphenyl)-3-(piperidin-1-yl)propan-1-one chloride
Synonyms: dyclonine
Molecular Weight (g/mol): 325.88
Molecular Formula: C18H28ClNO2
InChi Key: KNZADIMHVBBPOA-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCCCOC1=CC=C(C=C1)C(=O)CCN1CCCCC1
Certifications/Compliance 
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results