accessibility menu, dialog, popup

UserName

CH3ONHSHN

CAS RN 56-04-2

IUPAC Name: 6-methyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
Synonyms: methylthiouracil 甲硫氧嘧啶
Molecular Weight (g/mol): 142.18
Molecular Formula: C5H6N2OS
InChi Key: HWGBHCRJGXAGEU-UHFFFAOYSA-N
SMILES: CC1=CC(=O)NC(=S)N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (5)
  • (2)
Physical Form 
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)