accessibility menu, dialog, popup

UserName

CH3(R)OOCH3

CAS RN 938-94-3

IUPAC Name: (2R)-2-(4-methylphenyl)propanoate
Synonyms: (2R)-2-(4-methylphenyl)propanoate
Molecular Weight (g/mol): 163.2
Molecular Formula: C10H11O2
InChi Key: KDYOFXPLHVSIHS-MRVPVSSYSA-M
SMILES: C[C@@H](C([O-])=O)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results