accessibility menu, dialog, popup

UserName

SNNOOHOHOOOHOOSNNOOHOOHOOHOOSr

CAS RN 135459-87-9

IUPAC Name: strontium(2+) bis(2-{5-[bis(carboxymethyl)amino]-2-carboxy-4-cyanothiophen-3-yl}acetate)
Synonyms: strontium(2+) bis(2-{5-[bis(carboxymethyl)amino]-2-carboxy-4-cyanothiophen-3-yl}acetate)
Molecular Weight (g/mol): 770.16
Molecular Formula: C24H18N4O16S2Sr
InChi Key: KVJIIORUFXGTFN-UHFFFAOYSA-L
SMILES: [Sr++].OC(=O)CN(CC(O)=O)C1=C(C#N)C(CC([O-])=O)=C(S1)C(O)=O.OC(=O)CN(CC(O)=O)C1=C(C#N)C(CC([O-])=O)=C(S1)C(O)=O
Quantity 
  • (2)
  • (1)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results